Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113646 Supplier Invitrogen™ Supplier No. PA5113646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAWPPQPQPR QPPPPAPPAL QPPNGRGADE EVELEGLEPQ DLEASAGPAA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%.

This gene encodes a nuclear-localized protein that may be induced by p53 and regulates the cell cycle by inhibiting G1 to S phase progression. The encoded protein may interact with other cell cycle regulators.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZBTB10
Gene Accession No. Q96DT7
Gene Alias 4832414A18; AU021107; RINZF; RINZFC; ZBTB10; zinc finger and BTB domain containing 10; zinc finger and BTB domain-containing protein 10; Zinc finger protein RIN ZF; zinc finger protein RINZF
Gene Symbols ZBTB10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human ZBTB10 (aa 36-85).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65986
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.