Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180327
Description
ZBTB11 Polyclonal specifically detects ZBTB11 in Human samples. It is validated for Western Blot.Specifications
ZBTB11 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ13426, MGC133303, zinc finger and BTB domain containing 11, zinc finger and BTB domain-containing protein 11, ZNF913, ZNF-U69274 | |
Rabbit | |
Affinity purified | |
RUO | |
27107 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_055230 | |
ZBTB11 | |
Synthetic peptide directed towards the N terminal of human ZBTB11. Peptide sequence SSEESYRAILRYLTNEREPYAPGTEGNVKRKIRKAAACYVVRGGTLYYQR. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Chicken: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction