Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553302 Supplier Invitrogen™ Supplier No. PA553302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKGEVQTVAS TQDLRVQNGG TAPPVASSEG TTTSLPTELG DCEIVLLVNG ELPEAEQNGE VGRQPEPQVS SEAESALSSV GCIADSHPEM ESVDLITKNN QTELETSNNR ENNTVSNIHP KLS Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 58%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB11
Gene Accession No. O95625
Gene Alias 9230110G02Rik; ZBTB11; zinc finger and BTB domain containing 11; zinc finger and BTB domain-containing protein 11; ZNF913; ZNF-U69274
Gene Symbols ZBTB11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB11. Recombinant protein control fragment (Product #RP-91597).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.