Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZBTB11 |
---|---|
Concentration | 0.45 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4791
![]() ![]() |
Proteintech Group Inc
252151AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ZBTB11 may be involved in transcriptional regulation.Specifications
ZBTB11 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O95625 | |
27107, 271377 | |
ZBTB11 Fusion Protein Ag17000 | |
Primary | |
-20°C | |
ZBTB11 |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ZBTB11, ZNF U69274 | |
ZBTB11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title