Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP233480
Description
ZBTB12 Polyclonal specifically detects ZBTB12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZBTB12 | |
Polyclonal | |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Q9Y330 | |
ZBTB12 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GLGIGGSVGGHLGELAQSSVPPSTVAPPQGVVKACY | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C6orf46, chromosome 6 open reading frame 46, D6S59E, G10Bat9, NG35HLA-B-associated transcript 9, Protein G10, zinc finger and BTB domain containing 12, zinc finger and BTB domain-containing protein 12 | |
Rabbit | |
Affinity Purified | |
RUO | |
221527 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction