Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561451
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561451 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561451 Supplier Invitrogen™ Supplier No. PA561451
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLGIGGSVGG HLGELAQSSV PPSTVAPPQG VVKACY Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 89%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB12
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB12
Gene Accession No. Q9Y330
Gene Alias Bat9; Bat-9; BC020447; C6orf46; D6S59E; G10; HLA-B-associated transcript 9; Ng35; Ng35 pseudogene; Protein G10; ZBTB12; zinc finger and BTB domain containing 12; zinc finger and BTB domain-containing protein 12
Gene Symbols ZBTB12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB12. Recombinant protein control fragment (Product #RP-99552).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221527
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.