Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB14 Antibody, Novus Biologicals™
SDP

Catalog No. p-7109015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP174134 100 μL
NBP17413420 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP174134 Supplier Novus Biologicals Supplier No. NBP174134
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB14 Polyclonal specifically detects ZBTB14 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen ZBTB14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q08376
Gene Alias hZF5, ZBTB14MGC126126, ZF5zfp-5, Zfp-161, Zfp-5, Zinc finger and BTB domain-containing protein 14, zinc finger protein 161 homolog, zinc finger protein 161 homolog (mouse), Zinc finger protein 478, Zinc finger protein 5 homolog, zinc finger protein homologous to Zfp161 in mouse, ZNF478zfp-161
Gene Symbols ZBTB14
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the middle region of Zfp161. Immunizing peptide sequence DDDVEEIGDQDDSPSDDTVEGTPPSQEDGKSPTTTLRVQEAILKELGSEE.
Molecular Weight of Antigen 51 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7541
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.