Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568602 Supplier Invitrogen™ Supplier No. PA568602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

Transcriptional activator of the dopamine transporter (DAT), binding it's promoter at the consensus sequence 5'-CCTGCACAGTTCACGGA-3'. Binds to 5'-d(GCC)(n)-3' trinucleotide repeats in promoter regions and acts as a repressor of the FMR1 gene. Transcriptional repressor of MYC and thymidine kinase promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZBTB14
Gene Accession No. Q08376
Gene Alias b2b1982Clo; Zbtb14; ZF5; ZFP161; zfp-161; ZFP161 zinc finger protein; Zfp5; Zfp-5; zinc finger and BTB domain containing 14; zinc finger and BTB domain-containing protein 14; Zinc finger protein 161; zinc finger protein 161 homolog; Zinc finger protein 478; zinc finger protein 5; Zinc finger protein 5 homolog; zinc finger protein homologous to Zfp161 in mouse; ZNF478
Gene Symbols ZBTB14
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse ZBTB14.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22666
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.