Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB16, Mouse anti-Human, Clone: 5B3, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16321
Specifications
ZBTB16 | |
Monoclonal | |
Unconjugated | |
In ascites (0.03% sodium azide). | |
PLZF/ZNF145 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
5B3 | |
ELISA (1:10000) Immunocytochemistry (1:200-1:1000) Immunofluorescence (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
zinc finger and BTB domain containing 16 | |
ZBTB16 | |
Recombinant protein corresponding to human ZBTB16 from E. coli. | |
RUO | |
7704 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction