Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552208 Supplier Invitrogen™ Supplier No. PA552208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSPENVDDVL AVATFLQMQD IITACHALKS LAEPATSPGG NAEALATEGG DKRAKEEKVA TSTLSRLEQA GRSTPIGPSR DLKEERGGQA QSAASGAEQT EKADAPREPP PVELKPDPTS GMAAAEAEAA LSESSEQEME VE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.

Transcription factor that can function as an activator or repressor depending on its binding partners, and by targeting negative regulators of cell cycle progression. Plays a critical role in early lymphocyte development, where it is essential to prevent apoptosis in lymphoid precursors, allowing them to survive in response to IL7 and undergo proper lineage commitment. Has been shown to bind to the promoters of adenovirus major late protein and cyclin D1 and activate transcription. Required for early embryonic development during gastrulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB17
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB17
Gene Accession No. Q13105
Gene Alias AA589413; LP-1; Miz1; Miz-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; mZ13; pHZ-67; Polyomavirus late initiator promoter-binding protein; ZBTB17; Zfp100; Zfp-100; zinc finger and BTB domain containing 17; zinc finger and BTB domain-containing protein 17; Zinc finger protein 100; zinc finger protein 151; zinc finger protein 151 (pHZ-67); zinc finger protein 60; Zinc finger protein Z13; ZNF151; ZNF60
Gene Symbols ZBTB17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB17. Recombinant protein control fragment (Product #RP-100705).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7709
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.