Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564686
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4686 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4686 Supplier Bethyl Laboratories, Inc Supplier No. A303262AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

ZBTB2 is a member of the POK (POZ domain Krüppel-like zinc finger) family and plays an important role in cell cycle regulation. It is a transcription factor and a potent repressor of the ARF-HDM2-p53-p21 pathway. ZBTB2 represses the transcription of the p21, p53 and ARF genes and activates HDM2 gene. ZBTB2 and Sp1 compete with each other in binding to the GC-box 5/6 elements and the two p53 binding elements. ZBTB2 directly interacted with p53 via its zinc fingers, inhibiting p53 binding and repressing transcription activation by p53.

Specifications

Antigen ZBTB2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZBTB2
Gene Accession No. Q8N680
Gene Alias Zinc finger and BTB domain-containing protein 2; ZNF437; KIAA1483; ZBTB2
Gene Symbols Zbtb2
Host Species Rabbit
Immunogen Between 464 and 514
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381990, 57621
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.