Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596881 Supplier Invitrogen™ Supplier No. PA596881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILTAASILQI KTVIDECTRI VSQNVGDVFP GIQDSGQDTP RGTPESGTSG QSSDTESGYL QSHPQHSVDR IYSALYACSM QNGSGERSFY SGAVVSHHET ALGLPRDHHM EDPSWITRIH ERSQQMERYL STTPETTHCR KQPRPVRIQT LVGNIHIKQE MEDDYDYYGQ QRVQILERNE SEECTEDTDQ AEGTESEPKG ESFDSGVSSS IGTEPDSVEQ QFGPGAARDS QAEPTQPEQA AEAPAEGGPQ TNQLETGASS PERSNEVEMD STVITVSNSS DKSVLQQPSV NTSIGQPLPS T; Positive Samples: Jurkat, Mouse brain; Cellular Location: Nucleus.

This gene, which was initially designated as dendritic cell-derived BTB/POZ zinc finger (DPZF), belongs to a family of transcription factors with an N-terminal BTB/POZ domain and a C-terminal DNA-bindng zinc finger domain. The BTB/POZ domain is a hydrophobic region of approximately 120 aa which mediates association with other BTB/POZ domain-containing proteins. This gene acts as a transcriptional repressor and plays a role in many processes including neurogenesis, glucose homeostasis, and postnatal growth. Mutations in this gene have been associated with Primrose syndrome as well as the 3q13.31 microdeletion syndrome. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Feb 2017].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB20
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZBTB20
Gene Accession No. Q8K0L9, Q9HC78
Gene Alias 1300017A20Rik; 7330412A13Rik; A930017C21Rik; BTB/POZ domain zinc finger factor HOF; BTB/POZ zinc finger protein DPZF; D16Wsu73e; dendritic cell-derived BTB/POZ zinc finger; Dendritic-derived BTB/POZ zinc finger protein; DPZF; HOF; Oda8; ODA-8S; POZ/zinc finger transcription factor ODA-8; PRIMS; RGD1560387; Zbtb20; Zfp288; zinc finger and BTB domain containing 20; zinc finger and BTB domain-containing protein 20; Zinc finger protein 288; ZNF288
Gene Symbols ZBTB20
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human ZBTB20 (NP_0564573).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26137, 288105, 56490
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.