Learn More
Invitrogen™ ZBTB22 Polyclonal Antibody

Description
Predicted to react with mouse based on sequence homology.
Specifications
Specifications
| Antigen | ZBTB22 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ZBTB22 |
| Gene Accession No. | O15209 |
| Gene Alias | 1110008J20Rik; AI265210; AI415166; Bing1; fru; fruitless; Protein BING1; ZBTB22; ZBTB22A; Zfp297; zinc finger and BTB domain containing 22; zinc finger and BTB domain-containing protein 22; Zinc finger and BTB domain-containing protein 22A; zinc finger protein 297; ZNF297; ZNF297A |
| Gene Symbols | ZBTB22 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.