Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111190
Description
Immunogen sequence: SAVGSGERRG GGPVFPAPVV GSGGATSGKL LLEADELCDD GGDGRGAVVP GAGLRRPTYT PPSIMPQKHW VYVKRGGNCP AP.
May be involved in transcriptional regulation.
Specifications
| ZBTB22 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15209 | |
| ZBTB22 | |
| Recombinant Protein corresonding to Human ZBTB22. Recombinant protein control fragment (Product #RP-100045). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| ZBTB22 | |
| 1110008J20Rik; AI265210; AI415166; Bing1; fru; fruitless; Protein BING1; ZBTB22; ZBTB22A; Zfp297; zinc finger and BTB domain containing 22; zinc finger and BTB domain-containing protein 22; Zinc finger and BTB domain-containing protein 22A; zinc finger protein 297; ZNF297; ZNF297A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9278 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction