Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531622
Description
Recommended positive controls: ZBTB24 recombinant protein transfected 293T cell. Predicted reactivity: Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in BMP2-induced transcription.
Specifications
ZBTB24 | |
Polyclonal | |
Unconjugated | |
ZBTB24 | |
BC055367; Bif1; BMP-induced factor 1; bone morphogenetic protein-induced factor 1; brain specific protein 1; Brain-specific protein 1; Bsg1; ICF2; KIAA0441; PATZ2; POZ (BTB) and AT hook containing zinc finger 2; Zbtb24; zinc finger and BTB domain containing 24; zinc finger and BTB domain-containing protein 24; Zinc finger protein 450; ZNF450 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43167 | |
ZBTB24 | |
Recombinant protein fragment corresponding to a region within amino acids 121 and 363 of Human ZBTB24. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction