Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563010
Description
Immunogen sequence: IGLDDGTADQ QRACPATQAL EEHQKPPVSI KQERCDPESV ISQSHPSPSS EVTGPTFTEN SVKIHLCHYC GERFDSRSNL R Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.
May be involved in transcriptional regulation. Classified as a putative transcription factor; human homolog is a DNA binding protein with two distantly spaced zinc fingers.
Specifications
ZBTB25 | |
Polyclonal | |
Unconjugated | |
Zbtb25 | |
2810462M08Rik; 2900064P18Rik; AI842128; C14orf51; KUP; Zbtb25; Zfp50; Zfp-50; zinc finger and BTB domain containing 25; zinc finger and BTB domain-containing protein 25; Zinc finger protein 46; zinc finger protein 50; zinc finger protein KUP; ZNF46 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7597 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P24278 | |
Zbtb25 | |
Recombinant protein corresponding to Human ZBTB25. Recombinant protein control fragment (Product #RP-100906). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction