Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598246 Supplier Invitrogen™ Supplier No. PA598246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB25 Polyclonal Antibody for Western Blot, IHC (P), ELISA

May be involved in transcriptional regulation. Classified as a putative transcription factor; human homolog is a DNA binding protein with two distantly spaced zinc fingers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB25
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Zbtb25
Gene Accession No. P24278
Gene Alias 2810462M08Rik; 2900064P18Rik; AI842128; C14orf51; KUP; Zbtb25; Zfp50; Zfp-50; zinc finger and BTB domain containing 25; zinc finger and BTB domain-containing protein 25; Zinc finger protein 46; zinc finger protein 50; zinc finger protein KUP; ZNF46
Gene Symbols Zbtb25
Host Species Rabbit
Immunogen Recombinant Human Zinc finger and BTB domain-containing protein 25 protein (266-435AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7597
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.