Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598247
Description
ZBTB25 Polyclonal Antibody for Western Blot, IHC (P), ELISA
May be involved in transcriptional regulation. Classified as a putative transcription factor; human homolog is a DNA binding protein with two distantly spaced zinc fingers.
Specifications
ZBTB25 | |
Polyclonal | |
Unconjugated | |
Zbtb25 | |
2810462M08Rik; 2900064P18Rik; AI842128; C14orf51; KUP; Zbtb25; Zfp50; Zfp-50; zinc finger and BTB domain containing 25; zinc finger and BTB domain-containing protein 25; Zinc finger protein 46; zinc finger protein 50; zinc finger protein KUP; ZNF46 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7597 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24278 | |
Zbtb25 | |
Recombinant Human Zinc finger and BTB domain-containing protein 25 protein (266-435AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction