Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554410 Supplier Thermo Scientific Supplier No. PA554410
Add to Cart
Add to Cart

ZBTB26 Polyclonal Antibody

ZBTB26 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB26
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB26
Gene Accession No. Q9HCK0
Gene Alias A630026F21Rik; bioref; KIAA1572; ZBTB26; zinc finger and BTB domain containing 26; zinc finger and BTB domain-containing protein 26; Zinc finger protein 481; zinc finger protein 483; zinc finger protein Bioref; ZNF481
Gene Symbols ZBTB26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB26. Recombinant protein control fragment (Product #RP-93061).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57684
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.