Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530781 Supplier Invitrogen™ Supplier No. PA530781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG. Predicted reactivity: Mouse (96%), Rat (96%), Chicken (93%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB26
Applications Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZBTB26
Gene Accession No. Q9HCK0
Gene Alias A630026F21Rik; bioref; KIAA1572; ZBTB26; zinc finger and BTB domain containing 26; zinc finger and BTB domain-containing protein 26; Zinc finger protein 481; zinc finger protein 483; zinc finger protein Bioref; ZNF481
Gene Symbols ZBTB26
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZBTB26. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57684
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.