Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB3 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232140F
Description
ZBTB3 Polyclonal antibody specifically detects ZBTB3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ZBTB3 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
FLJ23392, zinc finger and BTB domain containing 3, zinc finger and BTB domain-containing protein 3 | |
The immunogen this antibody was made to, maps to a region between residue 524 to 574 of human Zinc Finger and BTB Domain Containing 3 using the numbering given in entry NP_079060.1 (GeneID 79842). | |
0.1 mL | |
Signal Transduction | |
79842 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction