Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521083
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521083 Supplier Invitrogen™ Supplier No. PA521083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-21083 can be used with blocking peptide PEP-1197.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB3, the related protein ZBTB2 is thought to be phosphorylated in response to the DNA damage, probably by either ATM or ATR. Other ZBTB proteins, such as ZBTB4 and ZBTB38 bind methylated DNA and repress transcription, suggesting that ZBTB3 may also act as a transcription repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ZBTB3
Gene Accession No. Q91X45, Q9H5J0
Gene Alias 4930563M09Rik; RGD1561140; ZBTB3; zinc finger and BTB domain containing 3; zinc finger and BTB domain-containing protein 3
Gene Symbols ZBTB3
Host Species Rabbit
Immunogen A 17 amino acid peptide near the carboxy terminus of human ZBTB3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499313, 75291, 79842
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.