Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557226 Supplier Invitrogen™ Supplier No. PA557226
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLREFSKWGV EASPGKAWER KRSLLRGAVG RYRGATGGDL FWAPFPSWGT MEFPEHSQQL Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB3, the related protein ZBTB2 is thought to be phosphorylated in response to the DNA damage, probably by either ATM or ATR. Other ZBTB proteins, such as ZBTB4 and ZBTB38 bind methylated DNA and repress transcription, suggesting that ZBTB3 may also act as a transcription repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB3
Gene Accession No. Q9H5J0
Gene Alias 4930563M09Rik; RGD1561140; ZBTB3; zinc finger and BTB domain containing 3; zinc finger and BTB domain-containing protein 3
Gene Symbols ZBTB3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB3. Recombinant protein control fragment (Product #RP-104215).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79842
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.