Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571052 Supplier Thermo Scientific Supplier No. PA571052
Add to Cart
Add to Cart

ZBTB34 Polyclonal Antibody

ZBTB34 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB34
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZBTB34
Gene Accession No. Q8NCN2
Gene Alias Gm349; KIAA1993; mKIAA1993; ZBTB34; zinc finger and BTB domain containing 34; zinc finger and BTB domain-containing protein 34; ZNF918
Gene Symbols ZBTB34
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human ZBTB34.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403341
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.