Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB37 Antibody (OTI1D7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274907AF405
Description
ZBTB37 Monoclonal specifically detects ZBTB37 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ZBTB37 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZBTB37 | |
Recombinant protein fragment corresponding to amino acids 131-361 of human ZBTB37 (NP_115911) produced in E.coli. | |
0.1 mL | |
84614.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1D7 | |
Western Blot | |
D430004I08Rik, MGC2629, zinc finger and BTB domain containing 37, zinc finger and BTB domain-containing protein 37, ZNF908 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction