Learn More
Invitrogen™ ZBTB38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593017
Description
Immunogen sequence: CMEMSEVFDD ASDQDSTDKP WRPYYNYKPK KKSRQLKKMR KVNWRKEHGN RSPSHKCKYP AELDCAVGKA PQDKPFEEEE TKEMPKLQCE LCDGDKAVGA GNQGRPHRHL TSRPYACELC AKQFQSPSTL KMHMRCHTGE KPYQCKTCGR CFSVQGNLQK HERIHLGLKE FVCQYCNKAF TLNETLKIHE RIHTGEKRYH CQFCFQRFLY LSTKRNHEQR HIREHNGKGY ACFQCPKICK TAAALGMHQK KHLFKSPSQQ EKIGDVCHEN SNPLENQHFI GSEDNDQKDN IQTGVENVVL; Positive Samples: Mouse heart, Mouse brain, Rat brain; Cellular Location: Chromosome, Nucleus.
ZBTB38 is a methyl CpG binding protein that binds to DNA sequences containing a single methylated CpG, thereby acting as a transcriptional repressor. It has been found to be highly expressed in the brain and may be involved in the differentiation of late postmitotic neurons.
Specifications
| ZBTB38 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB38 | |
| A930014K01Rik; CIBZ; CtBP-interacting BTB zinc finger protein; FLJ22332; FLJ31131; FLJ35036; PPP1R171; protein phosphatase 1, regulatory subunit 171; RGD1310136; si:ch73-160p19.2; UniProtKB:Q8BW71}; ZBTB38; zbtb38 {ECO:0000250; zenon; Zenon homolog; zinc finger and BTB domain containing 38; zinc finger and BTB domain-containing protein 38; Zinc finger protein expressed in neurons; zinc finger transcription factor Kaiso; ZNF921 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 253461 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NAP3 | |
| ZBTB38 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 896-1195 of human ZBTB38 (NP_0010738812). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.