Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566252
Description
Immunogen sequence: EHSYAVSSVAE AYRSQPVREH DGSSPGNTGK ENCEALAAKP KTCRKPKTFS IPQDSDSATE NIPPPPVSNL EVNQERSPQP AAVLTRSKSP NNEGDV Highest antigen sequence identity to the following orthologs - mouse 64%, rat 68%.
ZBTB38 is a methyl CpG binding protein that binds to DNA sequences containing a single methylated CpG, thereby acting as a transcriptional repressor. It has been found to be highly expressed in the brain and may be involved in the differentiation of late postmitotic neurons.
Specifications
| ZBTB38 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB38 | |
| A930014K01Rik; CIBZ; CtBP-interacting BTB zinc finger protein; FLJ22332; FLJ31131; FLJ35036; PPP1R171; protein phosphatase 1, regulatory subunit 171; RGD1310136; si:ch73-160p19.2; UniProtKB:Q8BW71}; ZBTB38; zbtb38 {ECO:0000250; zenon; Zenon homolog; zinc finger and BTB domain containing 38; zinc finger and BTB domain-containing protein 38; Zinc finger protein expressed in neurons; zinc finger transcription factor Kaiso; ZNF921 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 253461 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NAP3 | |
| ZBTB38 | |
| Recombinant Human ZBTB38. Recombinant protein control fragment (Product #RP-106931). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction