Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568564 Supplier Invitrogen™ Supplier No. PA568564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 91%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

ZBTB38 is a methyl CpG binding protein that binds to DNA sequences containing a single methylated CpG, thereby acting as a transcriptional repressor. It has been found to be highly expressed in the brain and may be involved in the differentiation of late postmitotic neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB38
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZBTB38
Gene Accession No. 0
Gene Alias A930014K01Rik; CIBZ; CtBP-interacting BTB zinc finger protein; FLJ22332; FLJ31131; FLJ35036; PPP1R171; protein phosphatase 1, regulatory subunit 171; RGD1310136; si:ch73-160p19.2; UniProtKB:Q8BW71}; ZBTB38; zbtb38 {ECO:0000250; zenon; Zenon homolog; zinc finger and BTB domain containing 38; zinc finger and BTB domain-containing protein 38; Zinc finger protein expressed in neurons; zinc finger transcription factor Kaiso; ZNF921
Gene Symbols ZBTB38
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZBTB38.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245007
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.