Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB38 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE134125UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1341-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1341-25UL Supplier MilliporeSigma Supplier No. ABE134125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-ZBTB38 Antibody is a highly specific rabbit polyclonal antibody that targets ZBTB38 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB38
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ZBTB38
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminus region of human Zinc finger and BTB domain-containing protein 38.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001073881
Test Specificity This rabbit polyclonal antibody specifically detects Zinc finger and BTB domain-containing protein 38 in human cells. It targets an epitope within 20 amino acids from the C-terminus region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.