Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553510 Supplier Invitrogen™ Supplier No. PA553510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MCETKFFTQW QLTLHRRDGI FENNIIVHPN DPLPGKLGLF SGAASPELKC AACGKVLAKD FHVVRGHILD HLNLKGQACS VCDQRHLNLC SLMWHTLSHL GISV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 92%.

ZBTB39 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB39
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB39
Gene Accession No. O15060
Gene Alias 7030401O21Rik; KIAA0352; mKIAA0352; RGD1306532; ZBTB39; zinc finger and BTB domain containing 39; zinc finger and BTB domain-containing protein 39; ZNF922
Gene Symbols ZBTB39
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB39. Recombinant protein control fragment (Product #RP-91691).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9880
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.