Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB39 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982989
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2989 Supplier Bioss Supplier No. BS13573R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB39 may be involved in transcriptional regulation.

Specifications

Antigen ZBTB39
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZBTB39
Gene Accession No. O15060
Gene Alias KIAA0352; ZBTB39; zinc finger and BTB domain containing 39; Zinc finger and BTB domain-containing protein 39; ZNF922
Gene Symbols ZBTB39
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZBTB39.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9880
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.