Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB4 (KAISO-L1) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE134225UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1342-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1342-25UG Supplier MilliporeSigma Supplier No. ABE134225UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-ZBTB4 (KAISO-L1) is a highly specific rabbit polyclonal antibody that targets Zinc finger and BTB domain-containing protein 4 and has been tested for use in Immunohistochemistry (Paraffin) and in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB4 (KAISO-L1)
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ZBTB4,KIAA1538
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 17 amino acids from the internal region of human Zinc finger and BTB domain-containing protein 4.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001122305
Test Specificity This rabbit polyclonal antibody detects Zinc finger and BTB domain-containing protein 4 (KAISO-L1) in human cells. It targets an epitope within 17 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.