Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534400
Description
A suggested positive control is Raji cell lysate. PA5-34400 can be used with blocking peptide PEP-1442.
May be involved in transcriptional regulation. [UniProt].
Specifications
| ZBTB40 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB40 | |
| BC059177; C230087D24; Gm571; KIAA0478; mKIAA0478; RGD1309866; ZBTB40; zinc finger and BTB domain containing 40; zinc finger and BTB domain-containing protein 40; ZNF923 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9923 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9NUA8 | |
| ZBTB40 | |
| A 19 amino acid synthetic peptide near the amino terminus of human ZBTB40. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction