Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB41 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP184119
Description
ZBTB41 Polyclonal specifically detects ZBTB41 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZBTB41 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
DKFZp686C06120, FLJ36199, FRBZ1, FRBZ1 protein (FRBZ1), zinc finger and BTB domain containing 41, zinc finger and BTB domain-containing protein 41 | |
Rabbit | |
Affinity Purified | |
RUO | |
360023 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZBTB41 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VECDQVTYTHSAGRPTPEALHCYQELPPSPDQRKLLSSLQYNKNLLKYLNDDRQKQPSFCDLLIIVEGKEFSAHKVVVAVGSSYFHACLSKNPSTDVVT | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction