Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551444 Supplier Invitrogen™ Supplier No. PA551444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGFLCDCTVL VGDARFPAHR AVLAACSVYF HLFYRDRPAG SRDTVRLNGD IVTAPAFGRL LDFMYEGRLD LRSLPVEDVL AAASYLHMYD IVKVCKGRLQ EKDRSLDPGN PAPGAEPAQP PC Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.

Transcriptional repressor. Specifically binds DNA and probably acts by recruiting chromatin remodeling multiprotein complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB42
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB42
Gene Accession No. B2RXF5
Gene Alias EG382639; Gm5188; LCCS6; protein simiRP58; simiRP58; ZBTB42; zinc finger and BTB domain containing 42; zinc finger and BTB domain-containing protein 42; ZNF925
Gene Symbols ZBTB42
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB42. Recombinant protein control fragment (Product #RP-88912).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100128927
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.