Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB43/ZNF297B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982993
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2993 Supplier Bioss Supplier No. BS13578R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF297B is a candidate transcription factor

Specifications

Antigen ZBTB43/ZNF297B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZBTB43
Gene Accession No. O43298
Gene Alias 1700010E06Rik; BOS_12004; KIAA0414; mKIAA0414; ortholog of zinc finger protein 297B ZNF297B; ZBTB22B; ZBTB43; Zfp297b; zinc finger and BTB domain containing 43; Zinc finger and BTB domain-containing protein 22B; Zinc finger and BTB domain-containing protein 43; Zinc finger protein 297B; ZNF297B; ZnF-x
Gene Symbols ZBTB43
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZBTB43/ZNF297B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23099
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.