Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562643 Supplier Invitrogen™ Supplier No. PA562643
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFGEYKHHMR VSRHIIRKPR IYECKTCGAM FTNSGNLIVH LRSLNHEASE LANYFQSSDF LVPDYLNQEQ EETLVQYDLG EHGFESNSSV QMPVISQYHS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 27%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB44
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB44
Gene Accession No. Q8NCP5
Gene Alias 6030404E16Rik; BTB (POZ) domain containing 15; BTB/POZ domain-containing protein 15; BTBD15; E230011F09; HSPC063; RGD1308984; Zbtb44; zinc finger and BTB domain containing 44; zinc finger and BTB domain-containing protein 44; Zinc finger protein 851; ZNF851
Gene Symbols ZBTB44
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB44. Recombinant protein control fragment (Product #RP-101596).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.