Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140176
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5140176 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5140176

Supplier: Invitrogen™ PA5140176

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: APPSFPDCAA GFLTAAADSA CEEPPAPTGL ADYSGAGRDF LRGAGSAEDV FPDSYVSTWH Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 78%.

ZNF499 may be involved in transcriptional regulation. In the central nervous system, it may play a role in glial cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB45
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB45
Gene Accession No. Q96K62
Gene Alias BB161562; Gm157; ZBTB45; Zfp499; zinc finger and BTB domain containing 45; zinc finger and BTB domain-containing protein 45; Zinc finger protein 499; ZNF499
Gene Symbols ZBTB45
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB45. Recombinant protein control fragment (Product #RP-109483).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84878
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.