Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB46 Antibody (PCRP-ZBTB46-2B8), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308298AF405
Description
ZBTB46 Monoclonal antibody specifically detects ZBTB46 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
ZBTB46 | |
Monoclonal | |
Alexa Fluor 405 | |
BTB (POZ) domain containing 4, BTB/POZ domain-containing protein 4, BTBD4, dJ583P15.7, dJ583P15.8, FLJ13502, RINZF, zinc finger and BTB domain containing 46, zinc finger and BTB domain-containing protein 46, Zinc finger protein 340, ZNF340 | |
Recombinant full-length human ZBTB46 protein (Uniprot: Q86UZ6) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PCRP-ZBTB46-2B8 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
140685 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction