Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB46 Polyclonal Antibody

Catalog No. PIPA536010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536010 Supplier Invitrogen™ Supplier No. PA536010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3. Predicted reactivity: Mouse (92%), Rat (91%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZBTB46 (Zinc Finger And BTB Domain Containing 46) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. ZBTB46 also functions as a transcriptional repressor for PRDM1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB46
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZBTB46
Gene Accession No. Q86UZ6
Gene Alias 2610019F01Rik; 4933406L05Rik; BTB (POZ) domain containing 4; BTB/POZ domain-containing protein 4; Btbd4; BTB-ZF protein expressed in effector lymphocytes; BZEL; dJ583P15.7; dJ583P15.8; RINZF; ZBTB46; zinc finger and BTB domain containing 46; zinc finger and BTB domain-containing protein 46; Zinc finger protein 340; ZNF340
Gene Symbols ZBTB46
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZBTB46. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140685
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.