Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144823
Description
Immunogen sequence: DLSVTEASSS DSRGERAELY AQVEEGLLGG EASYLGPPLT PEKDDALHQA TAVANLRAAL MSKNSLLSLK ADVLGDDGSL LFEYLPRGAH SLSLNEFTVI RKKFKCPYCS FSAMHQCILK R Antigen sequence identity: Mouse 93%; Rat 92%.
ZBTB46 (Zinc Finger And BTB Domain Containing 46) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. ZBTB46 also functions as a transcriptional repressor for PRDM1.
Specifications
ZBTB46 | |
Polyclonal | |
Unconjugated | |
ZBTB46 | |
2610019F01Rik; 4933406L05Rik; BTB (POZ) domain containing 4; BTB/POZ domain-containing protein 4; Btbd4; BTB-ZF protein expressed in effector lymphocytes; BZEL; dJ583P15.7; dJ583P15.8; RINZF; ZBTB46; zinc finger and BTB domain containing 46; zinc finger and BTB domain-containing protein 46; Zinc finger protein 340; ZNF340 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
140685 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UZ6 | |
ZBTB46 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 315-435. Recombinant protein control fragment (Product #RP-109726). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction