Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB48 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18174025UL
Description
ZBTB48 Polyclonal specifically detects ZBTB48 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZBTB48 | |
Polyclonal | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
GLI-Kruppel family member HKR3, HKR3zinc finger and BTB domain-containing protein 48, Krueppel-related zinc finger protein 3, Protein HKR3, zinc finger and BTB domain containing 48, Zinc finger protein 855, ZNF855pp9964 | |
Rabbit | |
Affinity Purified | |
RUO | |
3104 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZBTB48 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TSVGQAAGGQSGLGPPASQNVNSHVKEPAGLEEEEVSRTLGLVPRDQEPRGSHSPQRPQLHSPAQSEGPSSLCGKLKQAL | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction