Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556467
Description
Immunogen sequence: TSVGQAAGGQ SGLGPPASQN VNSHVKEPAG LEEEEVSRTL GLVPRDQEPR GSHSPQRPQL HSPAQSEGPS SLCGKLKQAL Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 59%.
ZBTB48 contains 1 BTB (POZ) domain and 11 C2H2-type zinc fingers. It belongs to the krueppel C2H2-type zinc-finger protein family and binds to and regulates the J and/or S elements in MHC II promoter.
Specifications
ZBTB48 | |
Polyclonal | |
Unconjugated | |
ZBTB48 | |
0610011D15Rik; AI327031; GLI-Krueppel family member HKR3; GLI-Kruppel family member HKR3; HKR3; Krueppel-related zinc finger protein 3; Krueppel-related zinc finger protein 3 homolog; pp9964; Telomere zinc finger-associated protein; Tzap; Zbtb48; zinc finger and BTB domain containing 48; zinc finger and BTB domain-containing protein 48; zinc finger and BTB domain-containing protein 48; telomere zinc finger-associated protein; zinc finger protein 855; ZNF855 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3104 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P10074 | |
ZBTB48 | |
Recombinant protein corresponding to Human ZBTB48. Recombinant protein control fragment (Product #RP-94580). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction