Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110589 Supplier Invitrogen™ Supplier No. PA5110589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDGSFVQHSV RVLQELNKQR EKGQYCDATL DVGGLVFKAH WSVLACCSHF FQSLYGDGSG GSVVLPAGFA EIFGLLLDFF YTGHLALTSG NRDQVLLAAR ELRVPEAVEL CQSFKPKTSV GQAAGGQSGL GPPASQNVNS HVKEPAGLEE EEVSRTLGLV PRDQEPRGSH SPQRPQLHSP AQSEGPSSLC GKLKQALKPC PLEDKKPEDC KVPPRPLEAE GAQLQGGSNE WEVVVQVEDD GDGDYMSEPE AVLTRRKSNV IRKPCAAEPA LSAGSLAAEP.

ZBTB48 contains 1 BTB (POZ) domain and 11 C2H2-type zinc fingers. It belongs to the krueppel C2H2-type zinc-finger protein family and binds to and regulates the J and/or S elements in MHC II promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB48
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZBTB48
Gene Accession No. P10074, Q1H9T6
Gene Alias 0610011D15Rik; AI327031; GLI-Krueppel family member HKR3; GLI-Kruppel family member HKR3; HKR3; Krueppel-related zinc finger protein 3; Krueppel-related zinc finger protein 3 homolog; pp9964; Telomere zinc finger-associated protein; Tzap; Zbtb48; zinc finger and BTB domain containing 48; zinc finger and BTB domain-containing protein 48; zinc finger and BTB domain-containing protein 48; telomere zinc finger-associated protein; zinc finger protein 855; ZNF855
Gene Symbols ZBTB48
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ZBTB48 (NP_005332.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100090, 3104
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.