Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109214
Description
Positive Control: Mouse spleen tissue lysate, HepG2, PC-3M, SH-SY5Y, human liver cancer tissue, human kidney tissue, mouse kidney tissue, rat kidney tissue.
ZBTB48 contains 1 BTB (POZ) domain and 11 C2H2-type zinc fingers. It belongs to the krueppel C2H2-type zinc-finger protein family and binds to and regulates the J and/or S elements in MHC II promoter.
Specifications
ZBTB48 | |
Polyclonal | |
Unconjugated | |
ZBTB48 | |
0610011D15Rik; AI327031; GLI-Krueppel family member HKR3; GLI-Kruppel family member HKR3; HKR3; Krueppel-related zinc finger protein 3; Krueppel-related zinc finger protein 3 homolog; pp9964; Telomere zinc finger-associated protein; Tzap; Zbtb48; zinc finger and BTB domain containing 48; zinc finger and BTB domain-containing protein 48; zinc finger and BTB domain-containing protein 48; telomere zinc finger-associated protein; zinc finger protein 855; ZNF855 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3104 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P10074 | |
ZBTB48 | |
Synthetic peptide within Human ZBTB48 aa 584-633. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction