Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB48 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA540455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540455 Supplier Thermo Scientific Supplier No. PA540455

Rabbit Polyclonal Antibody

Peptide sequence: MDGSFVQHSV RVLQELNKQR EKGQYCDATL DVGGLVFKAH WSVLACCSHF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%

ZBTB48 Polyclonal antibody specifically detects ZBTB48 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB48
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZBTB48
Gene Accession No. P10074
Gene Alias 0610011D15Rik; AI327031; GLI-Krueppel family member HKR3; GLI-Kruppel family member HKR3; HKR3; Krueppel-related zinc finger protein 3; Krueppel-related zinc finger protein 3 homolog; pp9964; Telomere zinc finger-associated protein; Tzap; Zbtb48; zinc finger and BTB domain containing 48; zinc finger and BTB domain-containing protein 48; zinc finger and BTB domain-containing protein 48; telomere zinc finger-associated protein; zinc finger protein 855; ZNF855
Gene Symbols ZBTB48
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZBTB48
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3104
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.