Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555081 Supplier Invitrogen™ Supplier No. PA555081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFSQYFRSLF QNSSSQKNDV FHLDVKNVSG IGQILDFMYT SHLDLNQDNI QVMLDTAQCL QVQNVLSLCH TFLKSATVVQ PPGMPCNSTL SLQSTLTPDA TCVISENYP Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.

Transcription factor. Inhibits cell proliferation by activating either CDKN1A/p21 transcription or RB1 transcription. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB49
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB49
Gene Accession No. Q6ZSB9
Gene Alias 4930518A03Rik; Zbtb49; Zfp509; zinc finger and BTB domain containing 49; zinc finger and BTB domain-containing protein 49; Zinc finger protein 509; ZNF509
Gene Symbols ZBTB49
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB49. Recombinant protein control fragment (Product #RP-93619).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166793, 305428
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.