Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554437 Supplier Invitrogen™ Supplier No. PA554437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEKEHMRVVV KSEPLSSPEP QDEVSDVTSQ AEGSESVEVE GVVVSAEKID LSPESSDRSF SDPQSSTDRV GDIHILEVTN NLEHKSTFSI SNFLNKSRGN NFTANQNN Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB5 was identified though sequence analysis as a POZ domain Kruppel-like zinc finger (POK) protein. Further experiments indicated that it binds DNA and can directly repress transcription of the cell cycle arrest gene p21. ZBTB5 can also interact with co-repressor histone deacetylase complexes such as BCoR, NCoR, and SMRT via its POZ domain, resulting in deacetylation of histones Ac-H3 and Ac-H4 at the proximal promoter. ZBTB5 stimulates both cell proliferation and cell cycle progression, suggesting that it may act as a potential proto-oncogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB5
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB5
Gene Accession No. O15062
Gene Alias 5930421I10; 9430083K24Rik; AI646847; expressed in brain and neural tube; Kiaa0354; mKIAA0354; RP11-397D12.5; Transcription factor ZNF-POZ; ZBTB5; zinc finger and BTB domain containing 5; zinc finger and BTB domain-containing protein 5
Gene Symbols ZBTB5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB5. Recombinant protein control fragment (Product #RP-93042).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9925
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.