Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562922
Description
Immunogen sequence: CTEPLSKYLE IDLSMKNNNQ HTDLCQSSDP DVKNEDENSD KDCEIIEISE DSPVNIDFHV K Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB6, also known as ZNF482, is highly expressed in brain. While little is known about ZBTB6, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB family member, ZBTB5, also acts as a transcriptional repressor, and can interact with co-repressor histone deacetylase complexes such as BCoR, NCoR, and SMRT via its POZ domain, suggesting that ZBTB6 may also act as a transcriptional repressor.
Specifications
ZBTB6 | |
Polyclonal | |
Unconjugated | |
ZBTB6 | |
A830092L04Rik; ZBTB6; Zfp482; ZID; zinc finger and BTB domain containing 6; zinc finger and BTB domain-containing protein 6; Zinc finger protein 482; zinc finger protein with interaction domain; ZNF482 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10773 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15916 | |
ZBTB6 | |
Recombinant protein corresponding to Human ZBTB6. Recombinant protein control fragment (Product #RP-101569). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction