Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112925
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112925 Supplier Invitrogen™ Supplier No. PA5112925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZBTB6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB6, also known as ZNF482, is highly expressed in brain. While little is known about ZBTB6, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB family member, ZBTB5, also acts as a transcriptional repressor, and can interact with co-repressor histone deacetylase complexes such as BCoR, NCoR, and SMRT via its POZ domain, suggesting that ZBTB6 may also act as a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZBTB6
Gene Accession No. Q15916
Gene Alias A830092L04Rik; ZBTB6; Zfp482; ZID; zinc finger and BTB domain containing 6; zinc finger and BTB domain-containing protein 6; Zinc finger protein 482; zinc finger protein with interaction domain; ZNF482
Gene Symbols ZBTB6
Host Species Rabbit
Immunogen Recombinant Human Zinc finger and BTB domain-containing protein 6 protein (115-424AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10773
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.